Cryogenic vials, glass
Leverancier: Hycultec
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9001-P 9011-P-Y 9002-0 9002-P 9022-P-W 9022-P-G 9022-P-R 9022-PL-W 9303-PP 9333-PP-W 9333-PP-Y 9333-PP-G 9333-PP-R 9333-PP-B 9333-PPL-R 9444-0-G 9404-PP 9444-PP-BLA 9444-PP-W 9333-0-G 9333-0-Y 9002-PP
SANB9001-PEA 0 EUR
SANB9001-P SANB9011-P-Y SANB9002-0 SANB9002-P SANB9022-P-W SANB9022-P-G SANB9022-P-R SANB9022-PL-W SANB9303-PP SANB9333-PP-W SANB9333-PP-Y SANB9333-PP-G SANB9333-PP-R SANB9333-PP-B SANB9333-PPL-R SANB9444-0-G SANB9404-PP SANB9444-PP-BLA SANB9444-PP-W SANB9333-0-G SANB9333-0-Y SANB9002-PP
Cryogenic vials, glass
flesjes
Glass vials, with screw cap. These vials are suitable for −20 °C deepfreezing. Screw caps with silicone stoppers make sure that the reagents are stored tight and secure.
  • Available with aluminium / coloured plastic / perforated plastic screw caps