Test sieves, 305×40 mm
Leverancier: Retsch GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60.159.000053 60.159.002000 60.159.000025 60.159.000032 60.159.000038 60.159.000045 60.159.000063 60.159.000075 60.159.000090 60.159.000106 60.159.000125 60.159.000150 60.159.000180 60.159.000212 60.159.000250 60.159.000300 60.159.000355 60.159.000425 60.159.000500 60.159.000600 60.159.000710 60.159.000850 60.159.001000 60.159.001180 60.159.001400 60.159.001700 60.159.002360 60.159.002800 60.159.003350 60.159.004000 60.159.004750 60.159.005600 60.159.006300 60.159.006700 60.159.008000 60.159.009500 60.159.011200 60.159.012500 60.159.013200 60.159.016000 60.159.019000 60.159.022400 60.159.025000 60.159.026500 60.159.031500 60.159.037500 60.159.045000 60.159.050000 60.159.053000 60.159.063000 60.159.075000 60.159.090000 60.159.100000 60.159.106000 60.159.125000
510-5505EA 250 EUR
510-5505 510-5526 510-5501 510-5502 510-5503 510-5504 510-5506 510-5507 510-5508 510-5509 510-5510 510-5511 510-5512 510-5513 510-5514 510-5515 510-5516 510-5517 510-5518 510-5519 510-5520 510-5521 510-5522 510-5523 510-5524 510-5525 510-5527 510-5528 510-5529 510-5530 510-5531 510-5532 510-5533 510-5534 510-5535 510-5536 510-5537 510-5538 510-5539 510-5540 510-5541 510-5542 510-5543 510-5544 510-5545 510-5546 510-5547 510-5548 510-5549 510-5550 510-5551 510-5552 510-5553 510-5554 510-5555
Test sieves, 305×40 mm
Zeven Testzeven
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieve meshes, frames and labeling comply with standards. The sieves are tested five times and are delivered with test report.
Certificaten: An individual inspection certificate for test materials monitoring according to ASTM E 11 is also available on request.
Bestelinformatie: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square. For a full overview, please go to website.