HPLC columns, LiChroCART®, Superspher® 60 RP-8e
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.50171.7140 1.50173.7140 1.50175.7140 1.50177.7140 1.50178.7140 1.50179.7140 1.50195.7140 1.50201.7140 1.50228.7140 1.50229.7140 1.50233.7140 1.50234.7140 1.50236.7140 1.50302.7140 1.51431.7140 1.51432.7140 1.51442.7140 1.51443.7140 1.51444.7140 1.51448.7140 1.50174.7140 1.50170.7140
1.50171.7140EA 0 EUR
1.50171.7140 1.50173.7140 1.50175.7140 1.50177.7140 1.50178.7140 1.50179.7140 1.50195.7140 1.50201.7140 1.50228.7140 1.50229.7140 1.50233.7140 1.50234.7140 1.50236.7140 1.50302.7140 1.51431.7140 1.51432.7140 1.51442.7140 1.51443.7140 1.51444.7140 1.51448.7140 1.50174.7140 1.50170.7140
HPLC columns, LiChroCART®, Superspher® 60 RP-8e
Kolommen voor chromatografie
Stainless steel LiChroCART® cartridges are available with various types of Superspher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.