Avantor® ACE® Excel® Silica, HPLC/UHPLC Columns, Analytical, 2 µm
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACE-1P7-GD2UEA 566 EUR
ACE-1P7-GD2U ACE-1P7-GD4U
Avantor® ACE® Excel® Silica, HPLC/UHPLC Columns, Analytical, 2 µm
Kolommen voor chromatografie
Avantor® ACE® Excel® Silica columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Wide range of particle sizes (2 to 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performanceMore Product Information