HPLC columns, LiChroCART®, Purospher® STAR RP-8e
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.51432.7220 1.50229.7194 1.50170.7220 1.50228.7194 1.50201.7194 1.50195.7220 1.50173.7220 1.50177.7194 1.50178.7194 1.51444.7194 1.50170.7194 1.50171.7194 1.51431.7220 1.51432.7194 1.50201.7220 1.50233.7220 1.50233.7194 1.50234.7194 1.50236.7194 1.50236.7220 1.50302.7194 1.50302.7220 1.51431.7194 1.51442.7194 1.51442.7220 1.51443.7194 1.51445.7194 1.50174.7194 1.50179.7194 1.51448.7194 1.51448.7220 1.50174.7220 1.50177.7220
1.51432.7220EA 827 EUR
1.51432.7220 1.50229.7194 1.50170.7220 1.50228.7194 1.50201.7194 1.50195.7220 1.50173.7220 1.50177.7194 1.50178.7194 1.51444.7194 1.50170.7194 1.50171.7194 1.51431.7220 1.51432.7194 1.50201.7220 1.50233.7220 1.50233.7194 1.50234.7194 1.50236.7194 1.50236.7220 1.50302.7194 1.50302.7220 1.51431.7194 1.51442.7194 1.51442.7220 1.51443.7194 1.51445.7194 1.50174.7194 1.50179.7194 1.51448.7194 1.51448.7220 1.50174.7220 1.50177.7220
HPLC columns, LiChroCART®, Purospher® STAR RP-8e
Kolommen voor chromatografie
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.