VWR®, Voorraadflessen, brede hals, rond, met schroefdoppen
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216-1708EA 38.6 EUR
216-1708 216-1730 216-1733 216-1919
VWR®, Voorraadflessen, brede hals, rond, met schroefdoppen
Flessen Voorraadflessen
Doorschijnend LDPE of PP met paarse schroefdoppen.
Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.
LDPE carboys
Ideal for high-purity applications, as they have high resistance to chemicals (widely used in ophthalmic products like eye droppers and nasal sprays).
Preferred resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impacts (will not break even if it falls down accidentally, whereas PP carboys may lead to cracking at negative temperatures)
LDPE product, will bounce back to their original shape as highly flexible material
Widely used in pharma industries
PP carboys
For sterile water storage, polypropylene (PP) carboys can be autoclaved before refilling to stop recurring bacterial growth.
Uitstekende chemische bestendigheid
Bedrijfstemperatuur van 0 tot 135 °C
Autoclaveerbaar
Certificaten: Hars voldoet aan US FDA 21 CFR, USP Klasse VI.
Opgelet: Polypropylene carboys are autoclavable. LDPE carboys are not autoclavable. Always remove the spigot and cap from the carboy before autoclaving.