Density hydrometers, M100
Leverancier: GERING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0345 0346 0347 0348 0349 0350 0351 0352 0344 0353 0354 0355
GERI0345EA 48.1 EUR
GERI0345 GERI0346 GERI0347 GERI0348 GERI0349 GERI0350 614-0115 614-0116 614-0193 GERI0353 GERI0354 GERI0355
Density hydrometers, M100
Hydrometers
Reference temperature for density measurements: 20 °C.
Certificaten: DIN 12791
Verpakking: In velvet lined case.