Film applicators, Bird-type
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VF1541 VF1542 VF1543 VF1544 VF1546 VF1571 VF1570 VF1572 VF1573 VF1574 VF1575 VF1576 VF1545 VF1540
SIMEVF1541EA 360 EUR
SIMEVF1541 SIMEVF1542 SIMEVF1543 SIMEVF1544 SIMEVF1546 SIMEVF1571 SIMEVF1570 SIMEVF1572 SIMEVF1573 SIMEVF1574 SIMEVF1575 SIMEVF1576 SIMEVF1545 SIMEVF1540
Film applicators, Bird-type
Filmapplicators Verf-/inktfilmapplicators
Bird-type film applicators feature one flat edge and one application side for applying a pre-defined coating thickness. Bird-type applicator single gap is available in film width 50 and 75 mm and can be customised for applying a host of different products onto flat and relatively solid substrates. All Bird-type film applicators are made from high grade stainless steel and are not affected by acid or base elements.
NOTE: The wet thickness that is obtained cannot be guaranteed. Wet thickness is dependent upon the solids and vehicle content of the wet material, as well as other factors. FilmMore Product Information