Dissolved oxygen bottles acc. to Winkler
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44267010 44267020
HECH44267010EA 16.7 EUR
HECH44267010 HECH44267020
Dissolved oxygen bottles acc. to Winkler
Flessen Flessen voor opgeloste zuurstof
Dissolved oxygen bottles acc. to Winkler, glass, clear, for the determination of oxygen dissolved in water.
  • Interchangeable plastic stopper
  • Frosted label
  • Precisely adjusted capacity
Order Now
Specificaties
Environmentally Preferable