Labels and tape for BBP®35 and BBP®37 thermal transfer benchtop printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
139950 145298 145300 145292 145293 143579 143580 145295 145294 143584 143581 145296 139949 139948 139947 139946 139945 139944 139943 139942 139941 139940 139939 139938 139937 139936 139935 139934 139932 139929 139928 143578 145297 143582
BRDY139950EA 324.66 EUR
BRDY139950 BRDY145298 BRDY145300 BRDY145292 BRDY145293 BRDY143579 BRDY143580 BRDY145295 BRDY145294 BRDY143584 BRDY143581 BRDY145296 BRDY139949 BRDY139948 BRDY139947 BRDY139946 BRDY139945 BRDY139944 BRDY139943 BRDY139942 BRDY139941 BRDY139940 BRDY139939 BRDY139938 BRDY139937 BRDY139936 BRDY139935 BRDY139934 BRDY139932 BRDY139929 BRDY139928 BRDY143578 BRDY145297 BRDY143582
Labels and tape for BBP®35 and BBP®37 thermal transfer benchtop printers
Etiketten
A variety of polyester and specialty materials for thermal transfer printing in wide format rolls designed for shape cutting multiple labels out of a single continuous roll.
B-423
This material is ideal for smooth-surface indoor labeling such as datacom equipment, patch panels, face plates, data ports and general industrial identification.
B-424
White matte paper labels with permanent rubber based adhesive for general purpose labelling.
B-428
Metallised polyester tape is ideal for indoor product labeling, asset tracking and general ID on smoother surfaces with a higher temperature limit.
B-432
Clear polyester tape is ideal for indoor labeling of smooth and slightly textured surfaces such as datacom equipment, faceplates, data ports, patch panels, and general industrial identification.
B-434 and B-486
Metallised polyester tape is ideal for indoor product labeling, asset tracking and general ID. B-434 is suitable for rough surfaces with a lower temperature limit. B-486 is suitable for very rough surfaces.
B-437
Topcoated polyvinylfluoride tape with a matte finish is designed for cable and wire bundle applications and label applications where self-extinguishing properties are required. Exhibits good solvent and heat resistance.
B-483
This material with a glossy finish is designed for high adhesion to textured metals and low surface energy plastics. For indoor use.
B-483/B-634
ToughStripe® floor marking features an ultra-aggressive adhesive which allows for printable messages to be placed on the floor. The floor marking roll is supplied with a separate roll of B-634 polyester overlaminate to protect your legend. Easy 3-step process: print label, position on floor, manually apply overlaminate. Withstands fork lift and pallet jack traffic without tearing or lifting.
B-484
Glossy white polyester tape with very aggressive adhesive ideal for angled and curved surface indoor labeling such as for panel labels and rating plates.