Bovine serum
Leverancier: Immunoreagents
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IMMRIR2420EA 102 EUR
IMMRIR2420 IMMRIR2421
Bovine serum
Serum
Normal bovine serum is used as a blocking agent or as a control for immunoassays. All serum is obtained from healthy, disease free animals. Serum is lipid extracted and buffer exchanged against phosphate buffer.
NOTE: All ImmunoReagents products are provided with a standard product CoA which includes application, purity and cross reactivity profiles. Additionally, products can by customised based on specific assay requirements, please contact your local Avantor sales office for more information.
Certificaten: (EC)142/2011 approved by APHIS, Reference Number NC-TEC-0002
Leveringsinformatie: Serum is supplied in lyophilised form without preservative. It is recommended to reconstitute the normal serum with DI water and add a preservative of choice or to aliquot reconstituted serum and store frozen to prevent contamination.