NMR tubes, PRECISION, thin walled, 5 mm Ø, SP Wilmad-LabGlass
Leverancier: SP Industries
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
507-PP-8QTZ-5 507-PP-7QTZ-5 528-PP-7QTZ-5 528-PP-8QTZ-5 535-PP-7QTZ-5 535-PP-8QTZ-5
WILM507-PP-8QTZ-5EA 402 EUR
WILM507-PP-8QTZ-5 WILM507-PP-7QTZ-5 WILM528-PP-7QTZ-5 WILM528-PP-8QTZ-5 WILM535-PP-7QTZ-5 WILM535-PP-8QTZ-5
NMR tubes, PRECISION, thin walled, 5 mm Ø, SP Wilmad-LabGlass
Buisjes NMR-buisjes
Quartz (natural quartz).
  • Possesses a low Boron density as below 0,1 ppm, which guarantees a clean background in Boron-11 NMR studies
  • 2× less dielectric constant than borosilicate glass, which helps improve the quality
  • Not recommended for EPR studies
  • Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6
Extremely low thermal expansion rate and high tensile strength. This feature makes quartz tubes 14 times more robust during cooling/heating process than corresponding Precision glass tubes. Naturally occurring quartz maintains over 85% transmission rate (10 mm thickness, with consideration of reflection loss) above 265 nm¹·², which makes quartz tubes preferable in photochemistry studies.