Rxi®-1ms Capillary Columns, (Fused Silica)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13302 13323 13353 13399 13354 43800 43801 43802 13326 13339 13324 13335 13337 13338
REST13302EA 678 EUR
REST13302 REST13323 REST13353 REST13399 REST13354 REST43800 REST43801 REST43802 REST13326 REST13339 REST13324 REST13335 REST13337 REST13338
Rxi®-1ms Capillary Columns, (Fused Silica)
Kolommen voor chromatografie
General purpose columns for arson accelerants, essential oils, hydrocarbons, pesticides, PCB congeners (e.g. Aroclor mixes), sulfur compounds, amines, solvent impurities, simulated distillation, oxygenates, gasoline range organics (GRO), refinery gases.
Non polar phase; Crossbond® 100% dimethyl polysiloxane
Temperature range: −60 to 330/350 °C
Similar phases: HP-1 ms, HP-1 msUI, DB-1 ms, DB-1 sUI, Ultra-1, VF-1 ms, ZB-1, ZB-1 ms.
Equivalent to USP G1, G2 and G38 phases.