Ultrasonic Test Sieve Cleaner
Leverancier: Nexopart
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HAVE205322259EA 4540 EUR
HAVE205322259 HAVE205323065 HAVE205322259UK HAVE205323065UK
Ultrasonic Test Sieve Cleaner
Ultrasoonbaden
It is used for thorough and gentle cleaning of sieves.
  • Easy handling and energy saving
  • Cleans up to five test sieves simultaneously
  • Functions well with USC cleaning fluid