Kruisbekers, borosilicaatglas
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632411006500_U 632411006940 632411006500
KAVA632411006500_UEA 30.8 EUR
KAVA632411006500_U KAVA632411006940 KAVA632411006500
Kruisbekers, borosilicaatglas
Bekers
Borosilicate glass, grade 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout.