Waterbaden, roestvrij staal, CORIO™
Leverancier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9903405 9903413 9903417 9903419 9903427 9903433 9903439
461-0921EA 302 EUR
461-0921 461-0922 461-0923 461-0924 461-0925 461-0926 461-0927
Waterbaden, roestvrij staal, CORIO™
Baden Waterbaden, ongeroerd
Stainless steel bath tanks in different sizes. For use with all CORIO C and CD circulators.
Temperature range: +20...+150 °C