Forceps set
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
000.120
232-0187EA 70.4 EUR
232-0187
Forceps set
Pincetten
Set contains four pairs of forceps made from stainless steel:
1× fine pointed
1× blunt ended
1× fine with guide pin
1× cover slip
Guidebook and case