Injection port liners, Uniliner®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20301 20305 20902 20903 20315 20301-214.1 20305-210.5 20308-214.1 20309-214.5 20316-205 20316-209.5 20316-213 20316-214.5 20316-216.5 20307 20308 20309 20314 20316 20319 20320
REST20301EA 0 EUR
REST20301 REST20305 REST20902 REST20903 REST20315 REST20301-214.1 REST20305-210.5 REST20308-214.1 REST20309-214.5 REST20316-205 REST20316-209.5 REST20316-213 REST20316-214.5 REST20316-216.5 REST20307 REST20308 REST20309 REST20314 REST20316 REST20319 REST20320
Injection port liners, Uniliner®
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
Liners are used for conversion of ¼" packed injection port.
  • Versatile, 0,53 mm int.Ø version can be used in the direct (DI) or on-column (OC) injection mode
  • Incorporates a gentle taper that seals the column and reduces dead volume in direct injection mode
  • Reduces solvent tailing