QuEChERS dSPE tubes, Q-sep™
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26123 26124 26125 26223 26224 26225 26215 26216 26217 26218 26126 26219 26220 26221 26222 26226 26242 26243 26244 26245
REST26123EA 228 EUR
REST26123 REST26124 REST26125 REST26223 REST26224 REST26225 REST26215 REST26216 REST26217 REST26218 REST26126 REST26219 REST26220 REST26221 REST26222 REST26226 REST26242 REST26243 REST26244 REST26245
QuEChERS dSPE tubes, Q-sep™
Kolommen voor chromatografie
QuEChERS dSPE tubes are used for extract cleanup. It is fast, simple sample Prep for multiresidue pesticide analysis. It has a advantage that no glassware required and it is ready to use. It can be individually labelled for easy sorbent identification and the tubes are preweighed with ultra pure sorbents.
Support original unbuffered, AOAC (2007.01), european (EN 15662) and mini-multiresidue QuEChERS methods.
Verpakking: Packaged in foil subpacks of 10 for enhanced protection and storage stability.