Diffraction/transmission gratings
Leverancier: 3B Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1003079 1003080
BINH1003079EA 0 EUR
BINH1003079 BINH1003080
Diffraction/transmission gratings
Educatief materiaal Educatief materiaal over fysica Leeractiviteiten voor optiek
Transmission grating for spectroscopic examinations and for experiments on diffraction and interference. Suitable to resolve the sodium D lines.
  • Mounted on glass carrier
  • Dimensions: 38×50 mm