Atomic absorption Spectrocopy (AAS) standards, multi-element in oil, Certipur®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.09479.0100 1.09469.0100
1.09479.0100EA 1160 EUR
1.09479.0100 1.09469.0100
Atomic absorption Spectrocopy (AAS) standards, multi-element in oil, Certipur®, Supelco®
Standaarden Kationische standaarden Standaarden met meerdere elementen
Standards dissolved in oil are ready-to-use calibration standards for the analysis of non-aqueous matrices.