Tapes for adhesion test
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP3007 SP3025 SP3020 SP3010
SIMESP3007EA 24.8 EUR
SIMESP3007 SIMESP3025 SIMESP3020 SIMESP3010
Tapes for adhesion test
Tape
This tape consists of 23 microns polyester films backing for improved moisture resistance, weight and space savings. These tapes are ideal to perform a cross cut hatch adhesion test.
  • Outstanding dielectric strength
  • Resistant to solvents, freon and transformer oils
  • Good peel strength to check paints and varnishes
  • Temperature resistance up to 130 °C, and classified under thermal class B
Different lengths available.
Colour: Transparent
Certificaten: Complies with ISO 2409:2003 or EN ISO 2409:1999.