Bulk material, Hydromatrix™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
198003 198004
VARI198003EA 329 EUR
VARI198003 VARI198004
Bulk material, Hydromatrix™
Sorbenten voor chromatografie Sorbenten voor SPE
SLE products use a broad-performance, inert diatomaceous earth sorbent for rapid, general sample preparation.