Sample dividers
Leverancier: ENDECOTTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1/2SSDSLOT
ENDE1/2SSDSLOTEA 0 EUR
ENDE1/2SSDSLOT
Sample dividers
Monsterverdelers
Endecotts hand held sample dividers will subdivide material samples into two smaller portions by a single pass. Further subdivisions can be attained by multiple passes. Each subdivision created retains the characteristics of the original sample.
  • Ideal for free flowing powders
  • Unit consists of divider plus 3 receiving boxes
  • Stainless steel with slot widths
  • Will subdivide material samples into two smaller portions by single pass
  • Each subdivision retains the characteristics of the original sample
  • Based on recommendations of BS 5309 and BS 3406/1