pH tester, Eutech EcoTestr pH 1 + pH 2
Leverancier: EUTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ECOPHTEST1 ECOPHTEST2
662-2014EA 83.5 EUR
662-2014 662-2015
pH tester, Eutech EcoTestr pH 1 + pH 2
Elektrochemische meters pH- en multiparametersystemen
This waterproof and accurate pH tester is ideal for quick pH measurements in hydroponic gardening, aquaculture, agriculture, pools, simple lab work and other water/wastewater applications. EcoTestr pH 2 features 3-point calibration and an ATC function.
  • Auto-calibration and auto-buffer recognition
  • Hold function freezes reading temporarily for ease of recording
  • IP 67 waterproof – tester floats on water for easy retrieval
  • Long sensor life – no need to change sensors
  • Click-lock battery compartment requires no tools for battery replacements
  • Transparent
More Product Information
Order Now
Specificaties
Model EcoTestr pH 1 EcoTestr pH 2
Environmentally Preferable
pH - bereik 0,0 - 14,0 0,0 - 14,0
pH - resolutie 0,1 0,1
pH - nauwkeurigheid ±0,1 ±0,1
pH - kalibratie 1 point (automatic) 3 points (automatic)
Temperatuurcompensatie - Automatic (ATC)
Aflezing 3½ digit LCD with annunciator 3½ digit LCD with annunciator
Voeding 4 × 1,5 V ‘A76’ micro alkaline batteries 4 × 1,5 V ‘A76’ micro alkaline batteries
Conformiteit IP67 IP67
b×d×h 163×45×30 mm 163×45×30 mm
Gewicht 0,09 kg 0,09 kg