Heat Barriers for CurveX Oven Data Loggers
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX2002 CX2005 CX2003 CX2004 CX2300 CX2017
SIMECX2002EA 2170 EUR
SIMECX2002 SIMECX2005 SIMECX2003 SIMECX2004 SIMECX2300 SIMECX2017
Heat Barriers for CurveX Oven Data Loggers
Dozen Opbergdozen
High quality insulation boxes with outer shield of high-grade stainless steel. The high grade stainless steel inner box is shielded with a carefully formulated mixture of insulation material, which makes the combination suitable to resist high temperatures for a long time.
  • Excellent logger protection against oven heat
  • Ferro plate for holding the magnet probes when not in use
  • Mounted cable hook allows the storage of surplus cable length