Inspection sets
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP5005 SP5000
SIMESP5005EA 874 EUR
SIMESP5005 SIMESP5000
Inspection sets
Coatinglaag testers
These basic sets are designed to perform all measurements and tests regarding painting.
Leveringsinformatie: Moisture meter, digital thermo/hygrometer, compass, illuminated surface microscope (to check the surface and paint layer), stainless steel cutter knife, pH-indicator paper (to check the acidity of concrete walls and floors), wedge (for checking reverse-side tolerance and thickness of glass), telescopic inspection mirror and lamp (for visual control), waterproof marker and MasterPaintPlate (multifunctional instrument to measure wet film thickness, cover of edges, thickness of glass, as well as providing an indication of the viscosity and adhesion of the coating).