Precision hydrometers, ASTM E100-10
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3222151 3222152 3222153 3222154 3222155 3222156 3222157 3222158 3222159 3222160
SCNL3222151EA 49.3 EUR
SCNL3222151 SCNL3222152 SCNL3222153 SCNL3222154 SCNL3222155 SCNL3222156 SCNL3222157 SCNL3222158 SCNL3222159 SCNL3222160
Precision hydrometers, ASTM E100-10
Hydrometers
Precision hydrometers (with or without thermometer) with reference temperature of 15 °C.
Error limit: ±0,5 kg/m³
Reference temperature: 15 °C
Certificaten: ASTM E100-10
Bestelinformatie: Available with a works or a DAkkS calibration certificate.
Verpakking: Each packed in a square plastic sleeve.