Hydrocarbon trap
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22013 22012
REST22013EA 245 EUR
REST22013 REST22012
Hydrocarbon trap
Filters Gasfilters
Hydrocarbon trap removes trace impurities from carrier gas. A 60 μm frit prevents gas contamination by purifier particles.
  • Ideal for purge and trap systems
  • Refillable and rechargeable
Maximum operating pressure: 1,25 bar
Maximum flow: 5 L/min
Dimensions: 235×57 mm