Pentaethylenehexamine mixture of isomers for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.40076.0025 8.40076.1000
8.40076.0025EA 33.9 EUR
8.40076.0025 8.40076.1000
Pentaethylenehexamine mixture of isomers for synthesis, Sigma-Aldrich®
Pentaethylenehexamine
Formule: C₁₀H₂₈N₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008167
CAS nummer: 4067-16-7
EINECS: 223-775-9
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Melting Point -35 °C
Molar Mass 232.37 g/mol
Boiling Point 380 °C
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 175 °C
Density 1.003 g/cm3 (20 °C)
pH 11 (10 g/l, H2O)
Ignition Point 360 °C
Melting point −35 °C
Vapour pressure -1 hPa (20 °C)
Assay of N 33 - 35%
Identity (IR) Passes test
Storage temperature +15...+25 °C