Precision saccharometers with different spans
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3281081 3281082 3281121
SCNL3281081EA 24.9 EUR
SCNL3281081 SCNL3281082 SCNL3281121
Precision saccharometers with different spans
Hydrometers
These precision saccharometers with different measuring spans are designed with or without thermometer.
  • Suitable for calibration
Saccharometers according to Brix and according to Balling are used to dertermine the sugar content. Degrees Brix or Balling are equivalent.
Reference temperature: 20 °C
Bestelinformatie: Available with a works or a DAkks calibration certificate on request.
Verpakking: In a square plastic sleeve.