Precision thermometers, for block thermostats, stem type
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
64304 64303
SCNL64304EA 46.5 EUR
SCNL64304 SCNL64303
Precision thermometers, for block thermostats, stem type
Thermometers Analoge thermometers
These thermometers are used for block thermostats, supplied in a plastic reaction tube, filled with 1 ml mineral oil.
  • Round, white back capillary form
  • Permanent graduations and lettering
  • Serial number and certificate (conformance and accuracy)
Blue filling, stem type.
Verpakking: Single packed in a cardboard box.