J.T.Baker® DILUT-IT™, Buffers, Acetate, Concentrated Solutions
Leverancier: Avantor
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D017-02EA
58.1
EUR
D017-02
D017-04
J.T.Baker® DILUT-IT™, Buffers, Acetate, Concentrated Solutions
Buffers
pH-buffers
Specially developed for dissolution testing of pharmaceutical dosage forms and drug discovery, easy to use DILUT-IT™ concentrates can reduce overall cost of ownership in the laboratory. Just add purified water and you are ready for testing.
- Produced from raw materials in accordance with USP guidelines in ISO-certified facilities
- Containers filled to ±0,5% of target fill volumes using specially designed filling equipment
- Application-optimised, function-tested for the challenges of dissolution testing
- Ideally suited for disintegration testing