Precision thermometers for block thermostats and water bath, stem type
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
64308 64306 64312 64314 64329 64310 64327 64325 64323 64321 64320 64318 64316
SCNL64308EA 38.5 EUR
SCNL64308 SCNL64306 SCNL64312 SCNL64314 SCNL64329 SCNL64310 SCNL64327 SCNL64325 SCNL64323 SCNL64321 SCNL64320 SCNL64318 SCNL64316
Precision thermometers for block thermostats and water bath, stem type
Thermometers Analoge thermometers
These thermometers are used for special applications.
  • Round, white or yellow back capillary form
  • Permanent graduation and lettering
  • Serial number
Red filling, stem type.
Verpakking: Single packed in plastic square outer covering.