Soil hydrometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3231032 3231031
SCNL3231032EA 55.2 EUR
SCNL3231032 SCNL3231031
Soil hydrometers
Hydrometers
Soil hydrometers according to the American Society for Testing Materials.
Reference temperature: 20 °C
Certificaten: ASTM E100-10.
Verpakking: In a square plastic sleeve.