Chemical resistant overalls, Tyvek® 200 Easysafe
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TSCHF5SWHLG010 TSCHF5SWHMD010 TSCHF5SWHXL010 TSCHF5SWH2X010 TSCHF5SWHSM010 TSCHF5SWH3X010
DUPOTSCHF5SWHLG010EA 739 EUR
DUPOTSCHF5SWHLG010 DUPOTSCHF5SWHMD010 DUPOTSCHF5SWHXL010 DUPOTSCHF5SWH2X010 DUPOTSCHF5SWHSM010 DUPOTSCHF5SWH3X010
Chemical resistant overalls, Tyvek® 200 Easysafe
Wegwerpkleding
These hooded overalls are made of Easysafe fabric, an optimised non woven PE fabric from DuPont. It is softer and with a more open structure than Tyvek®. It is therefore less protective than Tyvek® (L1431N), but offers adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable.
  • Robust yet lightweight (<180 g per garment)
  • Two-piece hood, zip closure with flap
  • Elasticated wrists, ankles and face
  • Elasticated waist (stitched-in)
  • Antistatic treatment (EN 1149-1) - on outside
  • Stitched external seams in yellow for visual identification and differentiation
Tyvek® 200 Easysafe overall offers demonstrably more protection than equivalent SMS garments and is more robust than microporous films.
Colour: White
Certificaten: Type 5/6, EN 1073-2, EN 1149-1; Cat. III
Verpakking: 100 per box, individually packed.