Jerricans, with self-sealing cap
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3941 3970 3943 3942
LAMA3941EA 5.3 EUR
LAMA3941 LAMA3970 LAMA3943 LAMA3942
Jerricans, with self-sealing cap
Flessen Jerrycans
HDPE, natural.
  • 1 and 2 L cans are provided inner-cap
  • Caps are autoclavable
Ideal for food storage.