Filtertips, ART® Ergonomic
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2330 2350 2330-05 2340-05
736-0208EA 0 EUR
736-0208 736-0210 613-3865 613-3866
Filtertips, ART® Ergonomic
Pipettips
The unique bi-fabricated wall of the ART® Ergonomic tip creates a strong, effective seal while requiring significantly lower tip insertion and tip ejection force. Available for Gilson Pipetman and FinnPipette Focus + Digital pipettes.
  • Reduce strains and injury
  • Secure and consistent seals
  • Less ejection force
  • Available in low retention polymer