Shoe covers, PP/CPE
Leverancier: Hubei Orient
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HOIT110490EA 184 EUR
HOIT110490 HOIT123605 114-9036 114-9035 114-9046 114-9045 114-9039 114-9038 114-9049 HOIT110017 114-9041 114-9040 HOIT110179 HOIT110024 HOIT110026
Shoe covers, PP/CPE
Wegwerpkleding
These non woven shoe covers are made of PP with a skid-resistant CPE coating on the sole.
  • High degree of sensitivity and high strength
  • Latex-free
  • Elasticated opening
Ideal for use in hospitals and medical environments.
Weight: 38 g/m² (PP fabric)