Assessment cabinets, Colorbox
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VF0600 VF1200
SIMEVF0600EA 2000 EUR
SIMEVF0600 SIMEVF1200
Assessment cabinets, Colorbox
Coatinglaag testers
The Colorbox is used to provide the illumination conditions for any visual inspection at a 45° angle.
  • Both interior and exterior finishes according to high quality standard
  • Multiple selectable light sources
  • Assessments of gloss, structure, damages and metamerism
Leveringsinformatie: Supplied with removable viewing table and counter.
Order Now
Specificaties
Environmentally Preferable
Hoek 45°