NMR sample tube washer
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.900.029 2.900.032
WITG2.900.029EA 217 EUR
WITG2.900.029 WITG2.900.032
NMR sample tube washer
Buisjes NMR-buisjes
Complete with 2 PTFE-adapters to fir 5 cuvets or NMR tubes of 5 and 10 mm diameter in different lengths. Vacuum operated washers allowing the individual washing of 1 to 5 tube, complete with caps and FEPcappilaries (box 1 pce.).