Hydrocarbon traps, capillary grade
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21991 21992
REST21991EA 227 EUR
REST21991 REST21992
Hydrocarbon traps, capillary grade
Filters Gasfilters
Packed with an extremely high surface area, baked coconut shell based activated carbon.
  • Purged with ultra high purity helium
  • Reduces organics to 0,1 ppm
  • Ready to use with any carrier gas including hydrogen
Maximum operating pressure: 17,2 bar