Interfaces
Leverancier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6915 31217 10019
HUBE6915EA 715 EUR
HUBE6915 HUBE31217 HUBE10019
Interfaces
Stopcontacten
Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue interface 0/4 to 20 mA or 0 to 10 V (bi-directional). Connections comply with NAMUR standard.