Protective and regenerating hand cream, Stokoderm® Aqua PURE
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24661._U 24668 24666 SAQ100ML SAQ1L
EVON24661._UEA 0 EUR
EVON24661._U 115-1476 EVON24666 115-2043 115-2338
Protective and regenerating hand cream, Stokoderm® Aqua PURE
Huidverzorging Huidcrèmes Handcrèmes
Aqua PURE is a barrier cream (W/O/W emulsion) that protects the skin when working with aqueous or water-miscible industrial substances and performing wet work. It has a repairing effect and stabilises, regenerates, and strengthens the skin barrier.
  • Contains bisabolol and witch hazel to ensure a high level of protection and care
  • Compatible with alcoholic hand disinfectants; does not impair the disinfection performance, in accordance with prEN 12054
  • Free from preservatives, perfume, colourants, and silicone
Suitable for use in industry, hospitals,More Product Information
Order Now
Specificaties
Environmentally Preferable