Regenerating hand cream, Stokoderm® Grip PURE
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SGP100ML SGP1L
115-1986EA 6.58 EUR
115-1986 115-2359
Regenerating hand cream, Stokoderm® Grip PURE
Huidverzorging Huidcrèmes Handcrèmes
Versatile skin protection product (O/W emulsion) for sensitive skin exposed to changing and non specific stresses. Can also be used under gloves. The product is effective for both purely oil-based and water-miscible or aqueous industrial substances. Ideal for activities in changing working areas.
  • Ingredients include an astringent active substance, wax-like agents and emollients
  • Free from classical preservatives, perfume, colourants and silicone
  • With gentle, moisture-binding ingredients
  • Can also be applied to the face due to good compatibility with mucosal