Regenerating hand and body cream, Stokolan® Sensitive PURE
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SSP100ML
115-2069EA 5.25 EUR
115-2069
Regenerating hand and body cream, Stokolan® Sensitive PURE
Huidverzorging Huidcrèmes Handcrèmes
Stokolan® Sensitive PURE is a regenerating care cream for particularly sensitive and exposed skin. Suitable for people prone to allergies and eczema.
  • Creatine supports cell renewal and builds up connective tissue
  • Compatible with alcoholic hand disinfectants; does not impair the disinfection performance, in accordance with prEN 12054
  • Colourant-free, silicone-free, perfume-free, preservative-free
This moisturising hand cream contains a high proportion of moisture-retaining, moisturising, and skin-conditioning ingredients, as well as the skin's own energy source, creatine, to support and strengthen the skin's defence system. It is suitable for use on the entire body, including the face and hands.
Certificaten: ECARF Quality Seal for allergy-friendly products, HACCP.