2,2'-[ethylenebis(oxy)]bisacetic acid for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 3,6-Dioxaoctanedioic acid
M-Clarity™ quality level = MQ200
8.43913.0050 8.43913.0250
8.43913.0050EA 0 EUR
8.43913.0050 8.43913.0250
2,2'-[ethylenebis(oxy)]bisacetic acid for synthesis, Sigma-Aldrich®
2,2'-[ethylenebis(oxy)]bisacetic acid
Formule: C₆H₁₀O₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00054502
CAS nummer: 23243-68-7
EINECS: 245-516-9
Order Now
Specification Test Results
Melting Point 72 °C
Molar Mass 178,14 g/mol
Hill Formula C6H10O6
HS Code 2918 99 90
Storage temperature 15...25 °C
Assay (HPLC, area%) ≥75%
Diglycolic acid (HPLC, area%) ≤5,0%
Water (K. F.) ≤20,0%
Identity (IR) passes test