Valves and loops, Sulfinert® treated
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20584 20585 20586
REST20584EA 2879 EUR
REST20584 REST20585 REST20586
Valves and loops, Sulfinert® treated
Ventielen Injectieventielen
Sulfinert® treated gas sample loops are ideal for samples containing low concentrations of sulphur and other active compounds.