DL-Tartaric acid for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (2R,3R)-rel-2,3-Dihydroxysuccinic acid, Racemic Acid
M-Clarity™ quality level = MQ200
8.43377.0100 8.43377.0500
8.43377.0100EA 41.2 EUR
8.43377.0100 8.43377.0500
DL-Tartaric acid for synthesis, Sigma-Aldrich®
DL-Tartaric acid
Formule: C₄H₆O₆
MW: 150.09 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00151254
CAS nummer: 133-37-9
EINECS: 205-105-7
Order Now
Specification Test Results
Solubility 215 g/l (25 °C)
Melting Point 198...204 °C
Molar Mass 150.09 g/mol
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 210 °C
pH 1.6 (100 g/l, H2O, 25 °C)
Ignition Point 425 °C
pH value 1,6 (100 g/l, H2O, 25 °C)
Vapor pressure - 0,1 hPa (20 °C)
Storage temperature 15...25 °C
Assay (acidimetric) ≥99%
Melting range (lower value) ≥198%
Melting range (upper value) ≤204 °C
Identity (IR) passes test