Bottles and tube racks
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HS120085 HS120086
HEATHS120085EA 37.9 EUR
HEATHS120085 HEATHS120086
Bottles and tube racks
Rekken Flessenrekken
Wire, epoxy coated.
  • Rack suitable for Bijou bottles and low profile tubes up to 25 mm diameter
  • Mesh bottom support
  • Chemically-resistant steel wire racks
For transporting and handling samples.